Learn More
Biomatik Corporation Human GNLY (Granulysin) ELISA Kit
Supplier:  Biomatik Corporation EKF58712
Encompass_Preferred
Human GNLY (Granulysin) ELISA Kit for detecting GNLY in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22749. Target Synonyms: GNLY; Granulysin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5986