Learn More
Biomatik Corporation Rat ApoC3 (Apolipoprotein C3) ELISA Kit
Supplier:  Biomatik Corporation EKF58720
Encompass_Preferred
Rat ApoC3 (Apolipoprotein C3) ELISA Kit for detecting ApoC3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 3.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06759. Target Synonyms: ApoC3; Apolipoprotein C3; APOC3; ApoC-III; MGC150353; Apo-CIII; APOCIII; apoC-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5994