Learn More
Biomatik Corporation Mouse ASK1 (Apoptosis Signal Regulating Kinase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58727
Encompass_Preferred
Mouse ASK1 (Apoptosis Signal Regulating Kinase 1) ELISA Kit for detecting ASK1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35099. Target Synonyms: ASK1; MAP3K5; MEKK5; Apoptosis signal-regulating kinase 1; ASK-1; ASK1MEKK 5; EC 2.7.11; MAP; ERK kinase kinase 5; MAPK; ERK kinase kinase 5; MEK kinase 5; MEKK5apoptosis signal regulating kinase 1; mitogen-activated protein kinase kinase kinase 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6000