Learn More
Biomatik Corporation Human LRP2 (Low-density lipoprotein receptor-related protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58729
Encompass_Preferred
Human LRP2 (Low-density lipoprotein receptor-related protein 2) ELISA Kit for detecting LRP2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P98164. Target Synonyms: LRP2; gp330; Megalin; LRP-2; Low-density lipoprotein receptor-related protein 2; Glycoprotein 330. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6002