Learn More
Biomatik Corporation Human Lipopolysaccharides, LPS ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34448
Encompass_Preferred
The Human Lipopolysaccharides, LPS ELISA Kit allows for quantitative determination of Lipopolysaccharides, LPS in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-8040