Learn More
Biomatik Corporation Human AGR2 (Anterior Gradient Protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58754
Encompass_Preferred
Human AGR2 (Anterior Gradient Protein 2) ELISA Kit for detecting AGR2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95994. Target Synonyms: AGR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6025