Learn More
Biomatik Corporation Human PAPPA (Pappalysin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58771
Encompass_Preferred
Human PAPPA (Pappalysin-1) ELISA Kit for detecting PAPPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40miu/ml. Sensitivity: 0.375miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13219. Target Synonyms: PAPP-A; Pappalysin-1; PAPPA; PAPA; DIPLA1; IGFBP-4ase; PAPPA1; ASBABP2; Pappalysin 1; Pregnancy Associated Plasma Protein A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6039