Learn More
Biomatik Corporation Human ACC (Acetyl-CoA Carboxylase) ELISA Kit
Supplier:  Biomatik Corporation EKF58775
Encompass_Preferred
Human ACC (Acetyl-CoA Carboxylase) ELISA Kit for detecting ACC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13085. Target Synonyms: ACC; Acetyl-CoA Carboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6043