Learn More
Biomatik Corporation Mouse Ache (Acetylcholinesterase) ELISA Kit
Supplier:  Biomatik Corporation EKF58779
Encompass_Preferred
Mouse Ache (Acetylcholinesterase) ELISA Kit for detecting Ache in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21836. Target Synonyms: Ache; Acetylcholinesterase; ACEE; ARACHE; N-ACHE; YT; Acetylcholinesterase YT blood group; Apoptosis related acetylcholinesterase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6046