Learn More
Biomatik Corporation Human CFH (Complement factor H) ELISA Kit
Supplier:  Biomatik Corporation EKF58782
Encompass_Preferred
Human CFH (Complement factor H) ELISA Kit for detecting CFH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 23.438-1500ng/ml. Sensitivity: 14.063ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08603. Target Synonyms: CFH; ARMD4; ARMS1; beta-1-H-globulin; CFH; CFHL3; FH; FHL1; HF1; HF2; HUS; adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; beta-1H; beta-1-H-globulin; CFHL3; complement factor H; factor H; factor H-like 1; FH; FHL1; H factor 1; H factor 1; complement; H factor 2; complement; HF; HF1ARMS1; HF2; HUSMGC88246. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6048