Learn More
Biomatik Corporation Human NFKB1 (Nuclear factor NF-kappa-B p105 subunit) ELISA Kit
Supplier:  Biomatik Corporation EKF58791
Encompass_Preferred
Human NFKB1 (Nuclear factor NF-kappa-B p105 subunit) ELISA Kit for detecting NFKB1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19838. Target Synonyms: NFKB1; NF-kB1; DNA-binding factor KBF1; EBP-1; NF-kappa-B; NFKB-p105; Nuclear factor NF-kappa-B p105 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; p105. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6057