Learn More
Biomatik Corporation Human Lipoxin A4, LXA4 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34450
Encompass_Preferred
The Human Lipoxin A4, LXA4 ELISA Kit allows for quantitative determination of Lipoxin A4, LXA4 in serum, plasma, cell culture supernates, urine, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 0.625 pg/mL-40 pg/mL. Sensitivity: 0.156 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-8042