Learn More
Biomatik Corporation Human ALAD (Delta-aminolevulinic acid dehydRatase) ELISA Kit
Supplier:  Biomatik Corporation EKF58796
Encompass_Preferred
Human ALAD (Delta-aminolevulinic acid dehydRatase) ELISA Kit for detecting ALAD in red blood cell lysis, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13716. Target Synonyms: ALAD; ALADH; Porphobilinogen synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6061