Learn More
Biomatik Corporation Human MTR (Methionine synthase) ELISA Kit
Supplier:  Biomatik Corporation EKF58798
Encompass_Preferred
Human MTR (Methionine synthase) ELISA Kit for detecting MTR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99707. Target Synonyms: MTR; Methionine synthase; 5-methyltetrahydrofolate--homocysteine methyltransferase; Vitamin-B12 dependent methionine synthase; MS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6063