Learn More
Biomatik Corporation Human CYP27B1 (25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF58803
Encompass_Preferred
Human CYP27B1 (25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial) ELISA Kit for detecting CYP27B1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O15528. Target Synonyms: CYP27B1; VD3 1A hydroxylase; Cytochrome P450VD1-alpha; Cytochrome P450C1 alpha; Cytochrome P450 subfamily XXVIIB polypeptide 1; Calcidiol 1-monooxygenase; Cytochrome p450 27B1; 25-OHD-1 alpha-hydroxylase; 25-hydroxyvitamin D; 3 1-alpha-hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6067