Learn More
Biomatik Corporation Human ST6GAL1 (Beta-galactoside alpha-2,6-sialyltransferase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58817
Encompass_Preferred
Human ST6GAL1 (Beta-galactoside alpha-2,6-sialyltransferase 1) ELISA Kit for detecting ST6GAL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15907. Target Synonyms: ST6GAL1; Alpha 2; 6-ST; B-cell antigen CD75; CD75; SIAT1; ST6N; B-cell antigen CD75; beta-galactoside alpha-2; 6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2; 6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2; 6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1; beta-galactoside alpha-2; 6-sialyltransferase; sialyltransferase 1; beta-galactoside alpha-2; 6-sialytransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6078