Learn More
Biomatik Corporation Rat STK11 (Serine/threonine-protein kinase STK11) ELISA Kit
Supplier:  Biomatik Corporation EKF58830
Encompass_Preferred
Rat STK11 (Serine/threonine-protein kinase STK11) ELISA Kit for detecting STK11 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: D4AE59. Target Synonyms: STK11; PJS; EC 2.7.11.1; LKB1 serine; threonine kinase 11; Peutz-Jeghers syndrome; PJS; PJS polarization-related protein LKB1; Renal carcinoma antigen NY-REN-19; serine; threonine kinase 11; serine; threonine-protein kinase 11; Serine; threonine-protein kinase LKB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6089