Learn More
Biomatik Corporation Mouse MYO (Myoglobin) ELISA Kit
Supplier:  Biomatik Corporation EKF58852
Encompass_Preferred
Mouse MYO (Myoglobin) ELISA Kit for detecting MYO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04247. Target Synonyms: Myo; Myoglobin; PVALB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6106