Learn More
Biomatik Corporation Rat Cartpt (Cocaine-and amphetamine-regulated transcript protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58856
Encompass_Preferred
Rat Cartpt (Cocaine-and amphetamine-regulated transcript protein) ELISA Kit for detecting Cartpt in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49192. Target Synonyms: Cartpt; CART; CART prepropeptide; cocaine and amphetamine regulated transcript; cocaine- and amphetamine-regulated transcript protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6109