Learn More
Biomatik Corporation Human TNFRSF10A (Tumor necrosis factor receptor superfamily member 10A) ELISA Kit
Supplier:  Biomatik Corporation EKF58860
Encompass_Preferred
Human TNFRSF10A (Tumor necrosis factor receptor superfamily member 10A) ELISA Kit for detecting TNFRSF10A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00220. Target Synonyms: TNFRSF10A; TRAIL R1; CD261; DR4; APO2; CD261 antigen; cytotoxic TRAIL receptor; Death receptor 4; DR4; DR4 TRAIL receptor 1; TNF-related apoptosis-inducing ligand receptor 1; TRAIL-R1; TRAILR-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6113