Learn More
Biomatik Corporation Rat AKR1B1 (Aldose reductase) ELISA Kit
Catalog No. 502296117
Encompass_Preferred
Rat AKR1B1 (Aldose reductase) ELISA Kit for detecting AKR1B1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07943. Target Synonyms: AKR1B10; Aldo-keto reductase family 1 member B10; Small intestine reductase; SI reductase; Aldose reductase-related protein; ARP; hARP; ARL-1; Aldose reductase-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-6117 Supplier Biomatik Corporation Supplier No. EKF58864
Rat AKR1B1 (Aldose reductase) ELISA Kit for detecting AKR1B1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07943. Target Synonyms: AKR1B10; Aldo-keto reductase family 1 member B10; Small intestine reductase; SI reductase; Aldose reductase-related protein; ARP; hARP; ARL-1; Aldose reductase-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.