Learn More
Biomatik Corporation Human Liver Type Fatty Acid Binding Protein (L-FABP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34454
Encompass_Preferred
The Human Liver Type Fatty Acid Binding Protein (L-FABP) ELISA Kit allows for quantitative determination of fatty acid binding protein 1, liver in serum, plasma and cell culture supernates. Species Reactivity: Human. Detection Range: 31.2 pg/mL-2000pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07148. Restriction: For Research Use Only.
Catalog No. 50-149-8044