Learn More
Biomatik Corporation Human SLC17A5 (Sialin) ELISA Kit
Supplier:  Biomatik Corporation EKF58875
Encompass_Preferred
Human SLC17A5 (Sialin) ELISA Kit for detecting SLC17A5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NRA2. Target Synonyms: SLC17A5; Sialin; AST; ISSD; NSD; SIASD; SLD; SD; Sodium; sialic acid cotransporter; Membrane glycoprotein HP59; sialic acid storage disease; Solute carrier family 17 member 5; solute carrier family 17; anion; sugar transporter; member 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6124