Learn More
Biomatik Corporation Mouse Serpina3c (Serine protease inhibitor A3C) ELISA Kit
Supplier:  Biomatik Corporation EKF58881
Encompass_Preferred
Mouse Serpina3c (Serine protease inhibitor A3C) ELISA Kit for detecting Serpina3c in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P29621. Target Synonyms: Serine protease inhibitor A3C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6128