Learn More
Biomatik Corporation Human BMP-6 (Bone morphogenetic protein 6) ELISA Kit
Supplier:  Biomatik Corporation EKF58891
Encompass_Preferred
Human BMP-6 (Bone morphogenetic protein 6) ELISA Kit for detecting BMP-6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22004. Target Synonyms: BMP-6; BMP6; VGR; VGR1; Vgr-1; bone morphogenetic protein 6; vegetal related growth factor; TGFB-related; vegetal-related; TGFB related cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6136