Learn More
Biomatik Corporation Human CD23 (Receptor II for the Fc Region of Immunoglobulin E) ELISA Kit
Supplier:  Biomatik Corporation EKF58893
Encompass_Preferred
Human CD23 (Receptor II for the Fc Region of Immunoglobulin E) ELISA Kit for detecting CD23 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06734. Target Synonyms: FcεRII; CD23; CLEC4J; Fc epsilon RII; FCER2; Fcer2a; IGEBF; Ly-42; BLAST-2; CD23A; CD23CD23 antigen; CLEC4JC-type lectin domain family 4 member J; C-type lectin domain family 4; member J; Fc fragment of IgE; low affinity II; receptor for; CD23; FCE2Fc fragment of IgE; low affinity II; receptor for; CD23A; fc-epsilon-RII; Fc-epsilon-RII; IGEBF; Immunoglobulin E-binding factor; low affinity immunoglobulin epsilon Fc receptor; Lymphocyte IgE receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6138