Learn More
Biomatik Corporation Mouse BMP-4 (Bone morphogenetic protein 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58903
Encompass_Preferred
Mouse BMP-4 (Bone morphogenetic protein 4) ELISA Kit for detecting BMP-4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21275. Target Synonyms: BMP-4; Bone Morphogenetic Protein 4; BMP2B; BMP2B1; MCOPS6; ZYME; DVR4; OFC11; ZYME. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6143