Learn More
Biomatik Corporation Mouse Fgf9 (Glia-activating factor) ELISA Kit
Supplier:  Biomatik Corporation EKF58912
Encompass_Preferred
Mouse Fgf9 (Glia-activating factor) ELISA Kit for detecting Fgf9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P54130. Target Synonyms: FGF9; FGF-9; GAF; HBFG-9; HBGF-9; MGC119914; MGC119915; SYNS3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6150