Learn More
Biomatik Corporation Human S1PR1 (Sphingosine 1-phosphate receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58915
Encompass_Preferred
Human S1PR1 (Sphingosine 1-phosphate receptor 1) ELISA Kit for detecting S1PR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21453. Target Synonyms: S1PR1; Sphingosine 1-phosphate receptor 1; S1P receptor 1; S1P1; Endothelial differentiation G-protein coupled receptor 1; Sphingosine 1-phosphate receptor Edg-1; S1P receptor Edg-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6153