Learn More
Biomatik Corporation Mouse Notch3 (Neurogenic locus notch homolog protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58930
Encompass_Preferred
Mouse Notch3 (Neurogenic locus notch homolog protein 3) ELISA Kit for detecting Notch3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61982. Target Synonyms: NOTCH3; CADASIL; CASILneurogenic locus notch homolog protein 3; Notch; Drosophila homolog 3; notch 3Notch homolog 3; Drosophila; Notch homolog 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6163