Learn More
Biomatik Corporation Rat PI (Proinsulin) ELISA Kit
Supplier:  Biomatik Corporation EKF58933
Encompass_Preferred
Rat PI (Proinsulin) ELISA Kit for detecting PI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01322. Target Synonyms: PI; Proinsulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6165