Learn More
Biomatik Corporation Mouse DAO (Diamine Oxidase) ELISA Kit
Supplier:  Biomatik Corporation EKF58941
Encompass_Preferred
Mouse DAO (Diamine Oxidase) ELISA Kit for detecting DAO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P18894. Target Synonyms: DAO; AOC1; DAO; Diamine Oxidase; Histaminase; KAO; ABP; amiloride binding protein 1; amine oxidase; copper-containing; Amiloride-binding protein; amiloride-sensitive amine oxidase; amiloride-sensitive amine oxidase [copper-containing]; AOC1DAOamiloride-binding protein-1; DAO1; Diamine oxidase; EC 1.4.3; EC 1.4.3.22; Histaminase; histaminase; KAO; Kidney amine oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6171