Learn More
Biomatik Corporation Rat VEGF-C (Vascular Endothelial Cell Growth Factor C) ELISA Kit
Supplier:  Biomatik Corporation EKF58944
Encompass_Preferred
Rat VEGF-C (Vascular Endothelial Cell Growth Factor C) ELISA Kit for detecting VEGF-C in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35757. Target Synonyms: VEGF-C; vascular endothelial growth factor C; Flt4 ligand; Flt4-L; Vascular endothelial growth factor-related protein; VRPFLT4 ligand DHM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6173