Learn More
Biomatik Corporation Mouse Acta2 (Actin, aortic smooth muscle) ELISA Kit
Supplier:  Biomatik Corporation EKF58948
Encompass_Preferred
Mouse Acta2 (Actin, aortic smooth muscle) ELISA Kit for detecting Acta2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P62737. Target Synonyms: α-SMA; Alpha-Smooth Muscle Actin; ACTA2; AAT6; Actin alpha 2; ACTSA; ACTVS; MYMY5; SMA; actin; alpha 2; smooth muscle; aorta; actin; aortic smooth muscle; alpha 2 actin; alpha SMA; alpha-actin-2; Alpha-actin-2; alpha-SMA; Cell growth-inhibiting gene 46 protein; growth-inhibiting gene 46. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6176