Learn More
Biomatik Corporation Rat SDF-1α (Stromal Cell Derived Factor 1α) ELISA Kit
Supplier:  Biomatik Corporation EKF58959
Encompass_Preferred
Rat SDF-1α (Stromal Cell Derived Factor 1α) ELISA Kit for detecting SDF-1α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9QZD1. Target Synonyms: SDF-1α; CXCL12 chemokine; C-X-C motif ligand 12; IRH; PBSF; SCYB12; SDF1; SDF-1; SDF1A; SDF1B; TLSF; TPAR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6184