Learn More
Biomatik Corporation Rat A2m (Alpha-2-macroglobulin) ELISA Kit
Supplier:  Biomatik Corporation EKF58962
Encompass_Preferred
Rat A2m (Alpha-2-macroglobulin) ELISA Kit for detecting A2m in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06238. Target Synonyms: A2m; Alpha-2-macroglobulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6187