Learn More
Biomatik Corporation Human PDX1 (Pancreas/duodenum homeobox protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58999
Encompass_Preferred
Human PDX1 (Pancreas/duodenum homeobox protein 1) ELISA Kit for detecting PDX1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P52945. Target Synonyms: PDX1; IDX1; IPF1; Glucose-sensitive factor; IDX-1GSF; Insulin promoter factor 1; insulin promoter factor 1; homeodomain transcription factor; Insulin upstream factor 1; IPF1pancreas; duodenum homeobox protein 1; Islet; duodenum homeobox-1; MODY4IUF1; pancreatic and duodenal homeobox 1; pancreatic-duodenal homeobox factor 1; PDX-1IPF-1; somatostatin transcription factor 1; Somatostatin-transactivating factor 1; STF-1IUF-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6218