Learn More
Biomatik Corporation Human MCAM (Cell surface glycoprotein MUC18) ELISA Kit
Supplier:  Biomatik Corporation EKF59028
Encompass_Preferred
Human MCAM (Cell surface glycoprotein MUC18) ELISA Kit for detecting MCAM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43121. Target Synonyms: MCAM; Melanoma Cell Adhesion Molecule; S-endo 1 endothelial-associated antigen; Melanoma-associated antigen MUC18; Melanoma-associated antigen A32; Cell surface glycoprotein P1H12; Melanoma cell adhesion molecule; MUC18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6242