Learn More
Biomatik Corporation Rat GP-ⅡbⅢa (Platelet Membrane Glycoprotein ⅡbⅢa) ELISA Kit
Supplier:  Biomatik Corporation EKF59035
Encompass_Preferred
Rat GP-ⅡbⅢa (Platelet Membrane Glycoprotein ⅡbⅢa) ELISA Kit for detecting GP-ⅡbⅢa in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8R2H2; D3ZAC0. Target Synonyms: GP-ⅡbⅢa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6245