Learn More
Biomatik Corporation Rat β-Hex A (β-hexosaminidase A) ELISA Kit
Supplier:  Biomatik Corporation EKF59037
Encompass_Preferred
Rat β-Hex A (β-hexosaminidase A) ELISA Kit for detecting β-Hex A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q641X3. Target Synonyms: β-Hex A; HEXA; TSD; beta-hexosaminidase subunit alpha; Beta-N-acetylhexosaminidase subunit alpha; hexosaminidase A; alpha polypeptide; Hexosaminidase subunit A; N-acetyl-beta-glucosaminidase subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6247