Learn More
Biomatik Corporation Human B2M (Beta2-microglobulin) ELISA Kit
Supplier:  Biomatik Corporation EKF59041
Encompass_Preferred
Human B2M (Beta2-microglobulin) ELISA Kit for detecting B2M in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P61769. Target Synonyms: BMG; β2-MG; Beta-2-Microglobulin; B2M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6251