Learn More
Biomatik Corporation Rat NTRK2 (Neurotrophic Tyrosine Kinase Receptor Type 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59058
Encompass_Preferred
Rat NTRK2 (Neurotrophic Tyrosine Kinase Receptor Type 2) ELISA Kit for detecting NTRK2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q63604. Target Synonyms: NTRK2; BDNF; NT-3 growth factors receptor; GP145-TrkB; neurotrophic tyrosine kinase; receptor; type 2; trk-B; Trk-B; TrkB tyrosine kinase; TRKBBDNF-tropomyosine receptor kinase B; tyrosine kinase receptor B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6265