Learn More
Biomatik Corporation Mouse Cspg4 (Chondroitin sulfate proteoglycan 4) ELISA Kit
Supplier:  Biomatik Corporation EKF59062
Encompass_Preferred
Mouse Cspg4 (Chondroitin sulfate proteoglycan 4) ELISA Kit for detecting Cspg4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8VHY0. Target Synonyms: Cspg4; MCSP; AN2; CSPG4; HMW-MAA; MCSPG; MEL-CSPG; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4; melanoma-associated; EC 3.6.3; HMW-MAA; MCSPChondroitin sulfate proteoglycan NG2; MCSPGMelanoma chondroitin sulfate proteoglycan; MEL-CSPG; MSK16Melanoma-associated chondroitin sulfate proteoglycan; NG2EC 2.7.8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6267