Learn More
Biomatik Corporation Mouse IDUA (α-L-iduronidase) ELISA Kit
Supplier:  Biomatik Corporation EKF59067
Encompass_Preferred
Mouse IDUA (α-L-iduronidase) ELISA Kit for detecting IDUA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48441. Target Synonyms: IDUA; IDUα; α-L-Iduronidase; IDUA; alpha ‑L‑Iduronidase; IDA; MPS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6272