Learn More
Biomatik Corporation Human ITGB2 (Integrin beta-2) ELISA Kit
Supplier:  Biomatik Corporation EKF59082
Encompass_Preferred
Human ITGB2 (Integrin beta-2) ELISA Kit for detecting ITGB2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05107. Target Synonyms: Cell surface adhesion glycoproteins LFA-1; CR3; p150; 95 subunit beta; Complement receptor C3 subunit beta; CD18; MFI7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6286