Learn More
Biomatik Corporation Human CNTN4 (Contactin 4) ELISA Kit
Supplier:  Biomatik Corporation EKF59087
Encompass_Preferred
Human CNTN4 (Contactin 4) ELISA Kit for detecting CNTN4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8IWV2. Target Synonyms: CNTN4; Brain-derived immunoglobulin superfamily protein 2; BIG-2; BIG-2; CNTN4; AXCAM; axonal-associated cell adhesion molecule; BIG-2Brain-derived immunoglobulin superfamily protein 2; contactin 4; contactin-4; neural cell adhesion protein BIG-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6289