Learn More
Biomatik Corporation Rat Avpr1a (Vasopressin V1a receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF59090
Encompass_Preferred
Rat Avpr1a (Vasopressin V1a receptor) ELISA Kit for detecting Avpr1a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30560. Target Synonyms: Avpr1a; AVPR V1a; V1aR; Antidiuretic hormone receptor 1a; arginine vasopressin receptor 1A; AVPR V1a; AVPR1; SCCL vasopressin subtype 1a receptor; V1a vasopressin receptor; V1aR; V1-vascular vasopressin receptor AVPR1A; Vascular; hepatic-type arginine vasopressin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6292