Learn More
Biomatik Corporation Rat ABCG2 (ATP-binding cassette sub-family G member 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59093
Encompass_Preferred
Rat ABCG2 (ATP-binding cassette sub-family G member 2) ELISA Kit for detecting ABCG2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q80W57. Target Synonyms: ABCG2; ATP Binding Cassette Transporter G2; ABCP; BCRP; CD338; MXR; ABC transporter; ABC15; ATP-binding cassette sub-family G member 2; ATP-binding cassette; sub-family G; WHITE; member 2; BCRP1; BCRPMRX; BMDP; Breast cancer resistance protein; CDw338; mitoxantrone resistance protein; Mitoxantrone resistance-associated protein; MXRMXR1; placenta specific MDR protein; Placenta-specific ATP-binding cassette transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6295