Learn More
Biomatik Corporation Human NUDT1 (7,8-dihydro-8-oxoguanine triphosphatase) ELISA Kit
Supplier:  Biomatik Corporation EKF59095
Encompass_Preferred
Human NUDT1 (7,8-dihydro-8-oxoguanine triphosphatase) ELISA Kit for detecting NUDT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P36639. Target Synonyms: NUDT1; MTH1; Nudix motif 1; Nudix motif 1; Nucleoside diphosphate-linked moiety X motif 1; 3.6.1.56; 8-oxo-dGTPase; Nucleoside diphosphate-linked moiety X motif 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6297