Learn More
Biomatik Corporation Rat LEPR (Leptin Receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF59112
Encompass_Preferred
Rat LEPR (Leptin Receptor) ELISA Kit for detecting LEPR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q62959. Target Synonyms: LEPR; Leptin R; Leptin receptor; CD295; LEP-R; LR; LEPRD; OB-R; OBR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6311